Aesculin sesquihydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6,7-Dihydroxycoumarin 6-glucoside sesquihydrate, Esculin sesquihydrate

M-Clarity™ quality level = MQ200

E8250-5G E8250-100G E8250-25G
SIALE8250-5GEA 58.3 GBP
SIALE8250-5G SIALE8250-100G SIALE8250-25G
Aesculin sesquihydrate, Sigma-Aldrich®
Aesculin sesquihydrate
Formula: C₁₅H₁₆O₉·1,5H₂O
MW: 367.31 g/mol
Melting Pt: 203…205 °C
MDL Number: MFCD00149492
CAS Number: 66778-17-4
EINECS: 208-517-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR