CytoFAST Elite Cytotoxic Safety Cabinets

Supplier: Faster
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CytoFAST
FASTFAS/1191/24EA 70380 GBP
FASTFAS/1191/24
CytoFAST Elite Cytotoxic Safety Cabinets
Furniture Storage Cabinets
CytoFAST Elite Cytotoxic Safety Cabinets belong to the latest generation of laminar airflow systems manufactured by Faster, in which the choice of materials of construction of the highest quality guarantees conformity to the strictest safety standards.

  • Ergonomic design
  • Real laminar airFlow
  • Innovative system of third bank H 14 HEPA/ULPA filtration
  • ECS® Eco Controling System
  • Anti bacterial coating
  • Inflatable gasketing
  • Silent operation
  • Protection grid
  • High level lighting
  • Easy handling and maintenance
  • Automatic safety service connections

They are triple HEPA filter technology cabinet, with 100% of the air filtered via the main H14 HEPA/ULPA filter directly below the work surface and then 70% of the air re-circulated via the recirculating H14 HEPA/ULPA filter within the cabinet, whilst the remaining 30% is discharged through an exhaust H14 HEPA/ULPA filter. CytoFAST Elite have been designed to guarantee the product, personnel and environmental protection and they are primary suitable for applications such as: preparation and handling of cytotoxic drugs, preparation and manipulation of antineoplastic chemotherapeutics, preparation and manipulation of CMR drugs and any other agents likely to cause genetic alterations or synergetic activities with other materials.

Since CytoFAST Elite meet also EN 12469 as for Class II Biohazard cabinets, they can be adopted also for a wide range of disciplines in applications such as: Microbiology, Virology, Haematology, Cell culture, Genetics, Handling of hazardous agents to human beings or animals.

Working aperture: 200 mm
Exhaust flow rate: 485 m³/h
Noise level: <55 dB(A)
Lighting level: >1200 lux
Electrical data: 230 V, 50 Hz
Current consumption: 3,6 A
Heat emission: 280 W
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR