Idex® Low-Pressure Union
Supplier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
02023-71 02023-70
COLE02023-71EA 0 GBP
COLE02023-71 COLE02023-70
Idex® Low-Pressure Union
Fittings and Connectors
Make solid connections in low-pressure applications.
  • Unions do not include fittings - provide own to match tubing size
  • Manufactured from inert polymers
  • 0,050" thru-hole diameter
  • PEEK body
  • 1 year warranty