Primeguard Plus Disposable Gowns
Supplier: LAB 3
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PG6L47WSST PG6M47WSST PG6S47WSST PG6XL47WSST PG6XXL47WSST
LBNOPG6L47WSSTEA 0 GBP
LBNOPG6L47WSST LBNOPG6M47WSST LBNOPG6S47WSST LBNOPG6XL47WSST LBNOPG6XXL47WSST
Primeguard Plus Disposable Gowns
Disposable Garments
Primeguard pro extra protective gown provides a barrier against fluids and contaminants while maintaining comfort and durability.
  • The unique laminated polypropylene structure incorporating polymer film and spunbond technology
  • Easy wear design with elasticated wrists and adjustable waistband makes this gown comfortable to wear
  • Optional thumb loop available for extra comfort and fit
  • Anti-static and low linting
  • Internal seams
Ideal for professionals needing reliable protection, it ensures safety and ease of movement, whilst the neck and waist ties ensure a secure fit.