RPE-Cy5-streptavidin conjugate
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16918 16919
AATB16918EA 122 GBP
AATB16918 AATB16919
RPE-Cy5-streptavidin conjugate
RPE-Cy5-streptavidin
RPE-Cy5-streptavidin conjugate comprises streptavidin (as the biotin-binding protein) with RPE-Cy5 covalently attached (as the fluorescent label).
  • Stable for 12 months
Streptavidin conjugates are widely used together with a biotin conjugate for specific detection of a variety of biological molecules since streptavidin has a very high binding affinity for biotin and its conjugates.
Caution: For Research Use Only