Anti-Annexin VI Mouse Monoclonal Antibody [clone: 4G3]

Supplier: ANTIBODIES.COM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A270554-100 A270554-50
ANTIA270554-100EA 345 GBP
ANTIA270554-100 ANTIA270554-50
Anti-Annexin VI Mouse Monoclonal Antibody [clone: 4G3]
Antibodies
Mouse monoclonal [4G3] antibody to Annexin VI for WB and ICC/IF with samples derived from Human, Rat and Mouse.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:1000 to 1:2000, ICC/IF: 1:1000

Type: Primary
Antigen: Annexin VI
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR