Thiomersal, EMPROVE® ESSENTIAL Ph. Eur., BP

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ500 (more info.)

8170430100 8170431000
SIAL8170430100EA 0 GBP
SIAL8170430100 SIAL8170431000
Thiomersal, EMPROVE® ESSENTIAL Ph. Eur., BP
Thiomersal

Thimerosal has strong antiseptic and antifungal capabilities and is therefore typically used as preservative in liquid dosage forms of cosmetics and pharmaceutical products.

Formula: 2-(C₂H₅HgS)C₆H₄CO₂Na
Melting Pt: 232...233 °C (dec.)
Flash Pt: 250 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00013062
CAS Number: 54-64-8
EINECS: 200-210-4
UN: 2025
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR