HPLC Columns, Hector M C8
Supplier: HICHROM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C8-M31001539 C8-M31001546 C8-M51001546
HISMC8-M31001539EA 654 GBP
HISMC8-M31001539 HISMC8-M31001546 HISMC8-M51001546
HPLC Columns, Hector M C8
Chromatography Columns
Hector M C8 is packed with alkylchain-group-bonded silica particles. It has a lower carbon load than Hector M C18.
  • Bonded phase: Octyl groups
It can be used for pharmaceutical, environmental, food, and other industrial chromatographic separations. It provides less retention and greater speed.