Ethylenetetracarbonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

T8809-25G T8809-5G
SIALT8809-25GEA 283 GBP
SIALT8809-25G SIALT8809-5G
Ethylenetetracarbonitrile, Sigma-Aldrich®
Ethylenetetracarbonitrile
Formula: C₆N₄
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 196…199 °C
Density: 1.35 g/cm³ (25 °C)
MDL Number: MFCD00001850
CAS Number: 670-54-2
EINECS: 211-578-0
UN: 3439
ADR: 6.1,II
Merck Index: 12,09336

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR