Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 532)

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

10192070 10192071
AATB10192070EA 328 GBP
AATB10192070 AATB10192071
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 532)
Antibodies
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR