Oil of cedar wood
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Cedar wood oil, Cedar Oil, Cedar Leaf Oil
38565.01
SERA38565.01EA
0
GBP
SERA38565.01
Oil of cedar wood
Oil of cedar wood
Density:
0.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00146327 CAS Number: 8000-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...