Sodium alginate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
180947-100G
180947-250G
180947-500G
SIAL180947-100GEA
53
GBP
SIAL180947-100G
SIAL180947-250G
SIAL180947-500G
Sodium alginate, Sigma-Aldrich®
Sodium alginate
Storage Temperature:
Ambient |
MDL Number:
MFCD00081310 CAS Number: 9005-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...