3-Thiophenecarbonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile
M-Clarity™ quality level = MQ100
597449-25G
597449-5G
SIAL597449-25GEA
139
GBP
SIAL597449-25G
SIAL597449-5G
3-Thiophenecarbonitrile, Sigma-Aldrich®
3-Thiophenecarbonitrile
Formula:
C₅H₃NS MW: 109.15 g/mol Boiling Pt: 82 °C (1013 hPa) Density: 1.1956 g/cm³ (20 °C) |
MDL Number:
MFCD00151852 CAS Number: 1641-09-4 EINECS: 216-687-7 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...