2-Thiophenecarbonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008056-25G 008056-100G
FLUO008056-25GEA 0 GBP
FLUO008056-25G FLUO008056-100G
2-Thiophenecarbonitrile
2-Thiophenecarbonitrile
Formula: C₅H₃NS
Boiling Pt: 190…192 °C (1013 hPa)
Density: 1.18 g/cm³ (15 °C)
MDL Number: MFCD00005416
CAS Number: 1003-31-2
EINECS: 213-706-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR