4-Fluorostyrene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Fluoro-4-vinylbenzene
001296-5G
001296-25G
FLUO001296-5GEA
0
GBP
FLUO001296-5G
FLUO001296-25G
4-Fluorostyrene
4-Fluorostyrene
Formula:
C₈H₇F MW: 122.14 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –34.5 °C Density: 1,022 g/cm³ (20 °C) |
MDL Number:
MFCD00000361 CAS Number: 405-99-2 EINECS: 206-975-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...