2-Fluorostyrene ≥98% stabilised
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Fluoro-2-vinylbenzene
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.10 %
A17425.06
A17425.22
A17425.06EA
85.1
GBP
A17425.06
A17425.22
2-Fluorostyrene ≥98% stabilised
2-Fluorostyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98%
CAS No.: 394-46-7
Molecular Formula: C8H7F
Formula Weight: 122.14
Storage Temperature: 2°C to 8°C
Flash Point: 35°(95°F)
Refractive Index: 1.5200
Odour: Recognizable
Boiling Point: 29-30°/4mm
Beilstein Reference: 1925226
UN No.: 1993
MDL No.: MFCD00013550
Formula:
C₈H₇F MW: 122.14 g/mol Boiling Pt: 29…30 °C (4 mmHg) Density: 1.020 Flash Pt: 35 °C (95 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013550 CAS Number: 394-46-7 EINECS: 206-896-1 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...