3,5-Di-tert-butylpyrocatechol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3,5-Di-tert-butylcatechol
008032-25G
008032-100G
FLUO008032-25GEA
0
GBP
FLUO008032-25G
FLUO008032-100G
3,5-Di-tert-butylpyrocatechol
3,5-Di-tert-butylpyrocatechol
Formula:
C₁₄H₂₂O₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 96…97 °C |
MDL Number:
MFCD00008819 CAS Number: 1020-31-1 EINECS: 213-816-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...