(±)-α-Methoxyphenylacetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid

M2876-10G M2876-25G M2876-5G
SIALM2876-10GEA 275 GBP
SIALM2876-10G SIALM2876-25G SIALM2876-5G
(±)-α-Methoxyphenylacetic acid, Sigma-Aldrich®
(±)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
Boiling Pt: 165 °C (24 hPa)
Melting Pt: 69…71 °C
MDL Number: MFCD00064217
CAS Number: 7021-09-2
EINECS: 230-300-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR