Heptafluorobutyric anhydride (HFAA)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
HFBA, 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, Perfluorobutyric anhydride, HFAA
001191.10G
001191.50G
001191-250G
FLUO001191.10GEA
0
GBP
FLUO001191.10G
FLUO001191.50G
FLUO001191-250G
Heptafluorobutyric anhydride (HFAA)
Heptafluorobutyric anhydride
Formula:
C₈F₁₄O₃ Boiling Pt: 108 °C (1013 hPa) Melting Pt: 14 °C Density: 1.653 g/cm³ (25 °C) |
MDL Number:
MFCD00000432 CAS Number: 336-59-4 EINECS: 206-410-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...