2-Fluoro-5-pyridineboronic acid 95%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid
L20387.03
L20387.06
L20387.MD
L20387.03EA
88.4
GBP
L20387.03
L20387.06
L20387.MD
2-Fluoro-5-pyridineboronic acid 95%
2-Fluoro-5-pyridineboronic acid
Purity: 95%
CAS No.: 351019-18-6
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Melting Point: 177-178° dec.
Beilstein Reference: 8974962
Formula:
C₅H₅BFNO₂ Melting Pt: 177…178 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03411559 CAS Number: 351019-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...