2-Bromo-5-pyridineboronic acid 95%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid
L20085.03
L20085.06
L20085.MD
L20085.03EA
95
GBP
L20085.03
L20085.06
L20085.MD
2-Bromo-5-pyridineboronic acid 95%
2-Bromo-5-pyridineboronic acid
Purity: 95%
CAS No.: 223463-14-7
Molecular Formula: C5H5BBrNO2
Formula Weight: 201.82
Melting Point: ca 165° dec.
Beilstein Reference: 9119918
Formula:
C₅H₅BBrNO₂ Melting Pt: ∼ 165 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03411558 CAS Number: 223463-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...