2-Hydroxyisobutyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydroxy-2-methylpropionic acid, α-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropanoic acid

M-Clarity™ quality level = MQ200

323594-100G 323594-25G
SIAL323594-100GEA 196 GBP
SIAL323594-100G SIAL323594-25G
2-Hydroxyisobutyric acid, Sigma-Aldrich®
2-Hydroxyisobutyric acid
Formula: C₄H₈O₃
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 77 °C
MDL Number: MFCD00004459
CAS Number: 594-61-6
EINECS: 209-848-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR