2-Fluoro-5-nitrobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF202.5
CFF202.25
006297-1G
006297-5G
006297-25G
CHMPCFF202.5EA
0
GBP
CHMPCFF202.5
CHMPCFF202.25
FLUO006297-1G
FLUO006297-5G
FLUO006297-25G
2-Fluoro-5-nitrobenzonitrile
2-Fluoro-5-nitrobenzonitrile
Formula:
C₇H₃FN₂O₂ MW: 166.11 g/mol Melting Pt: 79 °C |
MDL Number:
MFCD00042299 CAS Number: 17417-09-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...