2-Fluoro-5-iodobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013037-5G 013037-25G
FLUO013037-5GEA 0 GBP
FLUO013037-5G FLUO013037-25G
2-Fluoro-5-iodobenzonitrile
2-Fluoro-5-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
MDL Number: MFCD03094225
CAS Number: 351003-36-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR