4-Amino-3-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013035-5G 013035-25G
FLUO013035-5GEA 0 GBP
FLUO013035-5G FLUO013035-25G
4-Amino-3-fluorobenzonitrile
4-Amino-3-fluorobenzonitrile
Formula: C₇H₅FN₂
MDL Number: MFCD00055559
CAS Number: 63069-50-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR