2-Amino-5-bromobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024155-25G 024155-5G
FLUO024155-25GEA 0 GBP
FLUO024155-25G FLUO024155-5G
2-Amino-5-bromobenzonitrile
2-Amino-5-bromobenzonitrile
Formula: C₇H₅BrN₂
MDL Number: MFCD00158946
CAS Number: 39263-32-6
EINECS: 254-387-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR