2-Amino-4,5-dimethoxybenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Cyano-4,5-dimethoxyaniline
013112-5G
013112-25G
FLUO013112-5GEA
0
GBP
FLUO013112-5G
FLUO013112-25G
2-Amino-4,5-dimethoxybenzonitrile
2-Amino-4,5-dimethoxybenzonitrile
Formula:
C₉H₁₀N₂O₂ Boiling Pt: 334 °C (1013 hPa) Melting Pt: 156 °C |
MDL Number:
MFCD00058682 CAS Number: 26961-27-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...