2-Amino-6-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003982-25G 003982-100G
FLUO003982-25GEA 0 GBP
FLUO003982-25G FLUO003982-100G
2-Amino-6-fluorobenzonitrile
2-Amino-6-fluorobenzonitrile
Formula: C₇H₅FN₂
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 127 °C
MDL Number: MFCD00015477
CAS Number: 77326-36-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR