Centrifugal Evaporators, Genevac EZ-2 4.0
Supplier: FASTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G/1005/24B G/1015/24
FASTG/1005/24BEA 81800 GBP
FASTG/1005/24B FASTG/1015/24
Centrifugal Evaporators, Genevac EZ-2 4.0
Evaporators Centrifugal Evaporators
The latest SP Genevac EZ-2 4.0 series centrifugal evaporator for parallel sample evaporation has been redesigned with simplicity in mind and is based on our decades of expertise in evaporation science. A modern and compact space saving design to find space in the fume hood or on the lab bench, the EZ-2 is now even more powerful and versatile to streamline evaporation workflows and accelerate breakthroughs in life science research.
  • New touchscreen interface, featuring more pre-set methods, ensures effortless day-in, day-out use
  • Ergonomic redesign with the front facing SpeedTrap™ jar for easy visibility and safe removal/replacement
  • EXAL™ technology enables crystallisation studies such as polymorph screening for added versatility
  • Designed to reliably handle all common organic solvents and acids
  • High power lamp and software improvements enhance overall performance
  • Enhanced environmentally friendly features for a greener footprint
  • Low maintenance for worry free user operation
The cutting-edge design of the EZ-2 delivers many advantages when it comes to solvent evaporation requirements and fitted with advanced technologies such as the Dri-Pure® anti-bumping system to prevent sample cross-contamination, automatic end of run detection, and sample temperature control software for precise temperature control. With unrivalled versatility, this ensures effortless, fast, and safe sample evaporation to give total peace of mind.
Features and benefits:
Complete sample protection
Crystallisation
No bumping
Variety of sample holders
'Press and go' control
Compatible with wide range of solvents
Compact and user-friendly
Fast lyophilisation
Application: PROTACs, medicinal chemistry, oligo synthesis, synthetic biology, RNA therapeutics, proteins and peptides, post purification sample handling, environmental analysis, clinical and diagnostic testing, food and beverage, metabolism and toxicology studies, natural products research, parallel chemistry, oils, fuels and fats, crystallography, forensic.
EZ-2 4.0 Plus is recommended for solvents with boiling points up to 165 °C and ideal for evaporating HPLC fractions and nitric acid.
EZ-2 4.0 Elite is recommended for solvents with boiling points up to 220 °C and suitable for evaporating high boiling point solvents and lyophilising aqueous based samples.