Bis(3-fluorophenyl)disulphide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009629-1G 009629-5G 009629-25G
FLUO009629-1GEA 0 GBP
FLUO009629-1G FLUO009629-5G FLUO009629-25G
Bis(3-fluorophenyl)disulphide
Bis(3-fluorophenyl) disulphide
Formula: C₁₂H₈F₂S₂
MDL Number: MFCD01318104
CAS Number: 63930-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR