Sodium chlorodifluoroacetate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001061-100G
001061-25G
FLUO001061-100GEA
0
GBP
FLUO001061-100G
FLUO001061-25G
Sodium chlorodifluoroacetate
Sodium chlorodifluoroacetate
Formula:
C₂ClF₂NaO₂ Melting Pt: 193…195 °C |
MDL Number:
MFCD00064771 CAS Number: 1895-39-2 EINECS: 217-586-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...