(±)-Nipecotic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

013177-100G 013177-25G 013177-5G
FLUO013177-100GEA 0 GBP
FLUO013177-100G FLUO013177-25G FLUO013177-5G
(±)-Nipecotic acid
(±)-Nipecotic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00005992
CAS Number: 498-95-3
EINECS: 207-873-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR