(±)-δ-Undecalactone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: delta-Undecanolactone, (±)-Tetrahydro-6-hexyl-2H-pyran-2-one, 5-Hydroxyundecanoic acid delta-Lactone, (±)-Undecan-5-olide, Undecanoic δ-lactone, 6-Hexyltetrahydro-2H-pyran-2-one

M-Clarity™ quality level = MQ300

W329401-100G-K W329401-1KG-K W329401-5KG-K
SIALW329401-100G-KEA 95.5 GBP
SIALW329401-100G-K SIALW329401-1KG-K SIALW329401-5KG-K
(±)-δ-Undecalactone, Sigma-Aldrich®
(±)-δ-Undecalactone
Formula: C₁₁H₂₀O₂
MDL Number: MFCD00006650
CAS Number: 710-04-3
EINECS: 211-915-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR