(±)-δ-Tetradecalactone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Nonyltetrahydro-2H-pyran-2-one, (±)-Tetradecan-5-olide, δ-Tetradecalactone, (±)-Tetrahydro-6-nonyl-2H-pyran-2-one

M-Clarity™ quality level = MQ300

W359009-100G-K W359009-1KG-K W359009-5KG-K
SIALW359009-100G-KEA 126 GBP
SIALW359009-100G-K SIALW359009-1KG-K SIALW359009-5KG-K
(±)-δ-Tetradecalactone, Sigma-Aldrich®
(±)-δ-Tetradecalactone
Formula: C₁₄H₂₆O₂
MDL Number: MFCD00059809
CAS Number: 2721-22-4
EINECS: 220-334-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR