(±)-γ-Dodecalactone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(±)-Dihydro-5-octylfuran-2(3H)-one, gamma-Dodecalactone, 5-Octyldihydrofuran-2(3H)-one, gamma-Dodecanolactone, gamma-Laurolactone, (±)-Dodecan-4-olide
M-Clarity™ quality level = MQ100
W240001-100G-K
W240001-1KG-K
W240001-5KG-K
SIALW240001-100G-KEA
81.5
GBP
SIALW240001-100G-K
SIALW240001-1KG-K
SIALW240001-5KG-K
(±)-γ-Dodecalactone, Sigma-Aldrich®
(±)-γ-Dodecalactone
Formula:
C₁₂H₂₂O₂ MW: 198.31 g/mol |
MDL Number:
MFCD00036499 CAS Number: 2305-05-7 EINECS: 218-971-6 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...