4-Acetylphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

021606-25G 021606-5G
FLUO021606-25GEA 0 GBP
FLUO021606-25G FLUO021606-5G
4-Acetylphenylboronic acid
4-Acetylphenylboronic acid
Formula: C₈H₉BO₃
MDL Number: MFCD01074667
CAS Number: 149104-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR