5-Fluorocytosine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one

006215-1G 006215-5G 006215-100G 006215-25G
FLUO006215-1GEA 0 GBP
FLUO006215-1G FLUO006215-5G FLUO006215-100G FLUO006215-25G
5-Fluorocytosine
5-Fluorocytosine
Formula: C₄H₄FN₃O
MW: 129.09 g/mol
Boiling Pt: 298
Melting Pt: 165
MDL Number: MFCD00006035
CAS Number: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR