4-Aminonicotinic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
024507-1G
024507-5G
FLUO024507-1GEA
0
GBP
FLUO024507-1G
FLUO024507-5G
4-Aminonicotinic acid
4-Aminonicotinic acid
Formula:
C₆H₆N₂O₂ |
MDL Number:
MFCD00234183 CAS Number: 7418-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...