4-(Bromomethyl)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(Bromomethyl)phenyl)metaboric acid, (4-(Bromomethyl)phenyl)boronic acid, Dihydroxy(4-(bromomethyl)phenyl)borane, 4-Bromomethylphenylboronic acid, 4-(Bromomethyl)benzeneboronic acid

011020-1G 011020-5G
FLUO011020-1GEA 0 GBP
FLUO011020-1G FLUO011020-5G
4-(Bromomethyl)phenylboronic acid
4-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MDL Number: MFCD01318113
CAS Number: 68162-47-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR