3,5-Bis(trifluoromethyl)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005569-1G 005569-5G 005569-25G
FLUO005569-1GEA 0 GBP
FLUO005569-1G FLUO005569-5G FLUO005569-25G
3,5-Bis(trifluoromethyl)phenylboronic acid
3,5-Bis(trifluoromethyl)phenylboronic acid
Formula: C₈H₅BF₆O₂
MDL Number: MFCD00051850
CAS Number: 73852-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR