2-Phenylpiperidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
022075-1G
022075-5G
FLUO022075-1GEA
0
GBP
FLUO022075-1G
FLUO022075-5G
2-Phenylpiperidine
2-Phenylpiperidine
Formula:
C₁₁H₁₅N MW: 161.25 g/mol |
MDL Number:
MFCD00270125 CAS Number: 3466-80-6 EINECS: 222-422-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...