2-Tolylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Methylphenyl)metaboric acid, 2-Methylphenylboronic acid, 2-Methylbenzeneboronic acid, Dihydroxy(2-methylphenyl)borane, o-Tolylboronic acid

M-Clarity™ quality level = MQ100

393606-1G 393606-25G 393606-5G
SIAL393606-1GEA 38.8 GBP
SIAL393606-1G SIAL393606-25G SIAL393606-5G
2-Tolylboronic acid, Sigma-Aldrich®
2-Tolylboronic acid
Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 160…166 °C
MDL Number: MFCD00093526
CAS Number: 16419-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR