Iron(III) citrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

F3388-1KG F3388-250G
SIALF3388-1KGEA 305 GBP
SIALF3388-1KG SIALF3388-250G
Iron(III) citrate, Sigma-Aldrich®
Iron (III) citrate
Formula: C₆H₅FeO₇
CAS Number: 28633-45-6
EINECS: 249-117-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR