(±)-2-Bromopalmitic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromohexadecanoic acid, (±)-2-Bromohexadecanoic acid, (±)-2-Bromohexadecylic acid
238422-10G
238422-50G
SIAL238422-10GEA
50.9
GBP
SIAL238422-10G
SIAL238422-50G
(±)-2-Bromopalmitic acid, Sigma-Aldrich®
(±)-2-Bromopalmitic acid
Formula:
H₃C(CH₂)₁₃BrCHCOOH MW: 335.32 g/mol Boiling Pt: 390 °C (1013 hPa) Melting Pt: 53 °C |
CAS Number:
18263-25-7 EINECS: 242-137-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...