Nitrilotriacetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: NTA, 2,2',2''-Nitrilotriacetic acid

M-Clarity™ quality level = MQ200

N9877-100G N9877-1KG N9877-500G N9877-5G
SIALN9877-100GEA 40 GBP
SIALN9877-100G SIALN9877-1KG SIALN9877-500G SIALN9877-5G
Nitrilotriacetic acid, Sigma-Aldrich®
Nitrilotriacetic acid
Formula: C₆H₉NO₆
MW: 191.14 g/mol
Boiling Pt: 498
Melting Pt: 242
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004287
CAS Number: 139-13-9
EINECS: 205-355-7
UN: 2923
ADR: 8 II
Merck Index: 12,06675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR