4-Hydroxy-3-nitrobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018723-25G 018723-5G
FLUO018723-25GEA 0 GBP
FLUO018723-25G FLUO018723-5G
4-Hydroxy-3-nitrobenzyl alcohol
4-Hydroxy-3-nitrobenzyl alcohol
Formula: C₇H₇NO₄
Melting Pt: 122…123 °C
MDL Number: MFCD00024249
CAS Number: 41833-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR