4,4'-Isopropylidenediphenol, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Bisphenol A, BPA, 2,2-Bis(4-hydroxyphenyl)propane
239658-250G
239658-50G
SIAL239658-250GEA
297
GBP
SIAL239658-250G
239658-50G
4,4'-Isopropylidenediphenol, Supelco®
4,4'-Isopropylidenediphenol
Formula:
C₁₅H₁₆O₂ Boiling Pt: 401 °C (1013 hPa) Melting Pt: 155 °C Density: 1.205 g/cm³ (20 °C) Flash Pt: 227 °C |
MDL Number:
MFCD00002366 CAS Number: 80-05-7 EINECS: 201-245-8 Merck Index: 13,01296 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...