2,4-Bis(trifluoromethyl)benzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006020-1G 006020-5G 006020-10G
FLUO006020-1GEA 0 GBP
FLUO006020-1G FLUO006020-5G FLUO006020-10G
2,4-Bis(trifluoromethyl)benzyl alcohol
2,4-Bis(trifluoromethyl)benzyl alcohol
Formula: C₉H₆F₆O
MW: 244.14 g/mol
MDL Number: MFCD00042491
CAS Number: 143158-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR