2,3-Difluoro-p-cresol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005760-1G
005760-5G
FLUO005760-1GEA
0
GBP
FLUO005760-1G
FLUO005760-5G
2,3-Difluoro-p-cresol
2,3-Difluoro-p-cresol
Formula:
C₇H₆F₂O |
MDL Number:
MFCD01631651 CAS Number: 261763-45-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...