N,N'-Diethylethylenediamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
126942-25G
126942-5G
SIAL126942-25GEA
205
GBP
SIAL126942-25G
SIAL126942-5G
N,N'-Diethylethylenediamine, Sigma-Aldrich®
N,N'-Diethylethylenediamine
Formula:
C₆H₁₆N₂ MW: 116.21 g/mol Boiling Pt: 152…153 °C (1013 hPa) Melting Pt: –11 °C Density: 0.811 g/cm³ (20 °C) |
MDL Number:
MFCD00009033 CAS Number: 111-74-0 EINECS: 203-902-4 UN: 2685 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...